Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorophenyl isothiocyanate, 98%

Chemical Identifiers

Chemical Name or Material 4-Fluorophenyl isothiocyanate
CAS 1544-68-9
Molecular Formula C7H4FNS
Formula Weight 153.18
MDL Number MFCD00004809
UN Number UN2923
Beilstein 636596
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1556106

  SDS
alfa aesar™
A1556106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1556114

  SDS
alfa aesar™
A1556114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.18
Boiling Point 228°C
TSCA TSCA
Health Hazard 1 Danger
Flash Point 89°C (192°F)
Quantity 5g
CAS 1544-68-9
UN Number UN2923
Assay 98%
MDL Number MFCD00004809
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 636596
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Melting Point 24° to 28°C
Molecular Formula C7H4FNS
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Chemical Name or Material 4-Fluorophenyl isothiocyanate
RTECS Number NX8760000
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.