Learn More
4-Fluorophenyl isocyanide, 95%, Thermo Scientific™
$229.26 - $229.26
Chemical Identifiers
CAS | 24075-34-1 |
---|---|
Molecular Formula | C7H4FN |
Molecular Weight (g/mol) | 121.11 |
MDL Number | MFCD04117575 |
InChI Key | FEUHGXVGNQTPGQ-UHFFFAOYSA-N |
Synonym | 1-isocyano-4-fluorobenzene, 4-fluorophenyl isocyanide, 4-fluorophenylisocyanide, benzene, 1-fluoro-4-isocyano-9ci, benzene,1-fluoro-4-isocyano |
PubChem CID | 2734801 |
IUPAC Name | 1-fluoro-4-isocyanobenzene |
SMILES | FC1=CC=C(C=C1)[N+]#[C-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC372620010
|
Thermo Fisher Scientific
AC372620010 |
1g | Glass bottle |
Each for $229.26
|
|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
24075-34-1 | |
121.11 | |
FEUHGXVGNQTPGQ-UHFFFAOYSA-N | |
2734801 | |
FC1=CC=C(C=C1)[N+]#[C-] |
C7H4FN | |
MFCD04117575 | |
1-isocyano-4-fluorobenzene, 4-fluorophenyl isocyanide, 4-fluorophenylisocyanide, benzene, 1-fluoro-4-isocyano-9ci, benzene,1-fluoro-4-isocyano | |
1-fluoro-4-isocyanobenzene |
Specifications
24075-34-1 | |
100.0 | |
Undesignated | |
94% min. (GC) | |
C7H4FN | |
MFCD04117575 | |
1-isocyano-4-fluorobenzene, 4-fluorophenyl isocyanide, 4-fluorophenylisocyanide, benzene, 1-fluoro-4-isocyano-9ci, benzene,1-fluoro-4-isocyano | |
Solubility in water: insoluble | |
1.19 | |
1-fluoro-4-isocyanobenzene | |
2734801 | |
Liquid or Solid | |
4-Fluorophenyl isocyanide |
94.0 | |
1.1900g/mL | |
62°C to 68°C (20.0 hPa) | |
Authentic | |
FC6H4NC | |
Glass bottle | |
1g | |
FEUHGXVGNQTPGQ-UHFFFAOYSA-N | |
FC1=CC=C(C=C1)[N+]#[C-] | |
121.11 | |
121.11 | |
95% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
RUO – Research Use Only