Learn More
4-Fluorophenyl chloroformate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 308320010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentSpecifications
4-Fluorophenyl chloroformate | |
38377-38-7 | |
100.0 | |
76°C | |
98% | |
C7H4ClFO2 | |
ClCO2C6H4F | |
1g | |
4-fluorophenyl chloroformate, 4-fluorophenyl carbonochloridate, acmc-1ad3b, 4-fluorophenyl chlorocarbonate, 4-fluoro phenyl chloroformate, chloroformic acid p-fluorophenyl ester, carbonochloridic acid 4-fluorophenyl ester | |
C1=CC(=CC=C1OC(=O)Cl)F | |
174.56 | |
174.56 |
98% | |
97.5 | |
1.3290g/mL | |
Authentic | |
Glass bottle | |
1.4923 to 1.4943 | |
MFCD00013257 | |
1.329 | |
MSBGPEACXKBQSX-UHFFFAOYSA-N | |
(4-fluorophenyl) carbonochloridate | |
2733256 | |
98% |
Chemical Identifiers
38377-38-7 | |
174.56 | |
MSBGPEACXKBQSX-UHFFFAOYSA-N | |
2733256 | |
C1=CC(=CC=C1OC(=O)Cl)F |
C7H4ClFO2 | |
MFCD00013257 | |
4-fluorophenyl chloroformate, 4-fluorophenyl carbonochloridate, acmc-1ad3b, 4-fluorophenyl chlorocarbonate, 4-fluoro phenyl chloroformate, chloroformic acid p-fluorophenyl ester, carbonochloridic acid 4-fluorophenyl ester | |
(4-fluorophenyl) carbonochloridate |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It
GHS Signal Word: Danger
RUO â Research Use Only