Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorophenyl 2-thienyl ketone, 97%

$40.23 - $145.70

Chemical Identifiers

Chemical Name or Material 4-Fluorophenyl 2-thienyl ketone
CAS 579-49-7
Molecular Formula C11H7FOS
Formula Weight 206.23
MDL Number MFCD00044554
Beilstein 137007
Synonym 4-Fluorophenyl 2-thienyl methanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2553309

  SDS
alfa aesar™
B2553309
10g Each for $40.23

AAB2553318

  SDS
alfa aesar™
B2553318
50g Each for $145.70
Description & Specifications

Specifications

Formula Weight 206.23
Assay 97%
MDL Number MFCD00044554
Molecular Formula C11H7FOS
CAS 579-49-7
Melting Point 97° to 99°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 10g
Chemical Name or Material 4-Fluorophenyl 2-thienyl ketone
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 4-Fluorophenyl 2-thienyl methanone
Beilstein 137007
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.