Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorophenoxyacetic acid hydrazide, 98+%

$43.86 - $43.86

Chemical Identifiers

Chemical Name or Material 4-Fluorophenoxyacetic acid hydrazide
CAS 1737-62-8
Molecular Formula C8H9FN2O2
Formula Weight 184.17
MDL Number MFCD00082651
Beilstein 2526730
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1064903

  SDS
alfa aesar™
L1064903
1g Each for $43.86
Description & Specifications

Specifications

Formula Weight 184.17
Chemical Name or Material 4-Fluorophenoxyacetic acid hydrazide
Assay ≥98%
Molecular Formula C8H9FN2O2
MDL Number MFCD00082651
CAS 1737-62-8
Beilstein 2526730
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 124° to 126°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.