Please login to your online account to display your discounted pricing

4-Fluorophenoxyacetic acid, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluorophenoxyacetic acid
Name Note 99+%
CAS 405-79-8
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H7FO3
Linear Formula FC6H4OCH2CO2H
Formula Weight 170.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC225620050

  SDS
thermo fisher scientific
225620050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 170.14
Quantity 5g
Color White to light beige
EINECS Number 206-974-5
Melting Point 103° to 106°C
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Name Note 99+%
Assay Percent Range 99+%
MDL Number MFCD00004304
CAS 405-79-8
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Chemical Name or Material 4-Fluorophenoxyacetic acid
Molecular Formula C8H7FO3
Linear Formula FC6H4OCH2CO2H
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.