missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorophenoxyacetic acid, 98+%

$39.64 - $129.23

Chemical Identifiers

CAS 405-79-8
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00004304
InChI Key ZBIULCVFFJJYTN-UHFFFAOYSA-N
Synonym 4-fluorophenoxyacetic acid, 2-4-fluorophenoxy acetic acid, p-fluorophenoxyacetic acid, 4-fluorophenoxy acetic acid, acetic acid, 4-fluorophenoxy, p-fluorophenoxy acetic acid, para-fluorophenoxyacetic acid, 4-fluorophenoxy acetic aci, alpha-carboxy-4-fluoroanisole, 2-p-fluorophenoxy acetic acid
PubChem CID 67882
IUPAC Name 2-(4-fluorophenoxy)acetic acid
SMILES C1=CC(=CC=C1OCC(=O)O)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0121606 View Documents Alfa Aesar™
L0121606
5g Each for $39.64
 
AAL0121614 View Documents Alfa Aesar™
L0121614
25g Each for $129.23
 
Specifications

Chemical Identifiers

405-79-8
170.139
ZBIULCVFFJJYTN-UHFFFAOYSA-N
67882
C1=CC(=CC=C1OCC(=O)O)F
C8H7FO3
MFCD00004304
4-fluorophenoxyacetic acid, 2-4-fluorophenoxy acetic acid, p-fluorophenoxyacetic acid, 4-fluorophenoxy acetic acid, acetic acid, 4-fluorophenoxy, p-fluorophenoxy acetic acid, para-fluorophenoxyacetic acid, 4-fluorophenoxy acetic aci, alpha-carboxy-4-fluoroanisole, 2-p-fluorophenoxy acetic acid
2-(4-fluorophenoxy)acetic acid

Specifications

104°C to 105°C
405-79-8
MFCD00004304
4-fluorophenoxyacetic acid, 2-4-fluorophenoxy acetic acid, p-fluorophenoxyacetic acid, 4-fluorophenoxy acetic acid, acetic acid, 4-fluorophenoxy, p-fluorophenoxy acetic acid, para-fluorophenoxyacetic acid, 4-fluorophenoxy acetic aci, alpha-carboxy-4-fluoroanisole, 2-p-fluorophenoxy acetic acid
C1=CC(=CC=C1OCC(=O)O)F
170.139
170.14
4-Fluorophenoxyacetic acid
5g
C8H7FO3
2259128
ZBIULCVFFJJYTN-UHFFFAOYSA-N
2-(4-fluorophenoxy)acetic acid
67882
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning