Please login to your online account to display your discounted pricing

4-Fluorophenoxy-ethylbromide, 95%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  124161000

 View more versions of this product

Catalog No. AC124161000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid After Melting
Assay ≥94%
Formula Weight 219.05
Color Clear colorless to brownish
Quantity 100g
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 4-Fluorophenoxy-ethylbromide
Infrared Spectrum Authentic
Name Note 95%
Assay Percent Range 95%
Molecular Formula C8H8BrFO
Linear Formula FC6H4O(CH2)2Br
MDL Number MFCD00044739
CAS 332-48-9
Synonym 1-(2-Bromoethoxy)-4-fluorobenzene
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5300 to 1.5320 (20°C, 589nm)

Specifications

Chemical Name or Material 4-Fluorophenoxy-ethylbromide
Name Note 95%
CAS 332-48-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H8BrFO
Linear Formula FC6H4O(CH2)2Br
Formula Weight 219.05
MDL Number MFCD00044739
Synonym 1-(2-Bromoethoxy)-4-fluorobenzene
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents