missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorophenoxy-ethylbromide, 95%, ACROS Organics™

Chemical Identifiers

CAS 332-48-9
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8H8BrFO
Linear Formula FC6H4O(CH2)2Br
Formula Weight 219.05
MDL Number MFCD00044739
Synonym 1-(2-Bromoethoxy)-4-fluorobenzene
Chemical Name or Material 4-Fluorophenoxy-ethylbromide, 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC124161000 SDS Thermo Fisher Scientific
124161000
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC124160250 SDS Fisher Scientific Acros
124160250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

332-48-9
100.0
C8H8BrFO
219.05
1-(2-Bromoethoxy)-4-fluorobenzene
≥94%
94.0
95%
FC6H4O(CH2)2Br
MFCD00044739
4-Fluorophenoxy-ethylbromide, 95%

Specifications

Clear colorless to brownish
Liquid After Melting
94.0
1-(2-Bromoethoxy)-4-fluorobenzene
1.5300 to 1.5320 (20°C, 589nm)
FC6H4O(CH2)2Br
332-48-9
4-Fluorophenoxy-ethylbromide, 95%
219.05
100g
100.0
Authentic
95%
C8H8BrFO
MFCD00044739
≥94%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.