missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluorophenol 99.0+%, TCI America™
Supplier: TCI America F0160500G
Specifications
4-Fluorophenol | |
47°C | |
C6H5FO | |
500 g | |
RHMPLDJJXGPMEX-UHFFFAOYSA-N | |
4-fluorophenol | |
9732 | |
≥99.0% (GC) |
371-41-5 | |
185°C | |
MFCD00002316 | |
p-fluorophenol, phenol, 4-fluoro, phenol, p-fluoro, para-fluorophenol, 4-fluoro phenol, 4-fluoro-phenol, ccris 665, 4-fluoranylphenol, fpn | |
OC1=CC=C(F)C=C1 | |
112.10 | |
112.10 | |
Crystalline Powder |
Chemical Identifiers
371-41-5 | |
112.10 | |
RHMPLDJJXGPMEX-UHFFFAOYSA-N | |
9732 | |
OC1=CC=C(F)C=C1 |
C6H5FO | |
MFCD00002316 | |
p-fluorophenol, phenol, 4-fluoro, phenol, p-fluoro, para-fluorophenol, 4-fluoro phenol, 4-fluoro-phenol, ccris 665, 4-fluoranylphenol, fpn | |
4-fluorophenol |
Safety and Handling
EINECSNumber : (3)-4229
RTECSNumber : SL4550000
TSCA : Yes
Recommended Storage : Refrigerator