Please login to your online account to display your discounted pricing

4-Fluorophenethylealcohol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluorophenethyl alcohol
CAS 7589-27-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9FO
Linear Formula FC6H4CH2CH2OH
Formula Weight 140.16
MDL Number MFCD00002898
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC311980250

  SDS
thermo fisher scientific
311980250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 140.16
Physical Form Liquid
Quantity 25g
Color Clear colorless
Density 1.1200g/mL
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.5060 to 1.5080 (20°C, 589nm)
Linear Formula FC6H4CH2CH2OH
Chemical Name or Material 4-Fluorophenethyl alcohol
CAS 7589-27-7
Assay Percent Range 97%
Molecular Formula C8H9FO
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
MDL Number MFCD00002898
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.