Please login to your online account to display your discounted pricing

4-Fluorophenethylamine, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  350950250

 View more versions of this product

Catalog No. AC350950250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 139.17
Quantity 25g
Color Clear colorless to slightly yellow
Density 1.061g/mL
CAS Max % 100.0
Chemical Name or Material 4-Fluorophenethylamine
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H10FN
Linear Formula FC6H4CH2CH2NH2
MDL Number MFCD00134208
CAS 1583-88-6
CAS Min % 98.5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.5060 to 1.5080 (20°C, 589nm)

Specifications

Chemical Name or Material 4-Fluorophenethylamine
Name Note 99%
CAS 1583-88-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H10FN
Linear Formula FC6H4CH2CH2NH2
Formula Weight 139.17
MDL Number MFCD00134208
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents