Please login to your online account to display your discounted pricing

4-Fluoroisoindoline hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluoroisoindoline hydrochloride
CAS 924305-06-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8FN·ClH
Formula Weight 173.62
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC435860010

 
thermo fisher scientific
AC435860010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0%
Formula Weight 173.62
Quantity 1g
Color White to off-white
Water 5% Max. (K.F.)
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Fluoroisoindoline hydrochloride
Assay Percent Range 97%
Molecular Formula C8H8FN·ClH
CAS 924305-06-6
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.