Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoroisobutyrophenone, 97%

$248.62 - $688.59

Chemical Identifiers

Chemical Name or Material 4'-Fluoroisobutyrophenone
CAS 26393-91-9
Molecular Formula C10H11FO
Formula Weight 166.19
MDL Number MFCD04038305
Synonym 1-(4-Fluorophenyl)-2-methyl-1-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2603603

  SDS
alfa aesar™
H2603603
1g Each for $248.62

AAH2603606

  SDS
alfa aesar™
H2603606
5g Each for $688.59
Description & Specifications

Specifications

Formula Weight 166.19
Assay 97%
MDL Number MFCD04038305
Molecular Formula C10H11FO
CAS 26393-91-9
Synonym 1-(4-Fluorophenyl)-2-methyl-1-propanone
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4'-Fluoroisobutyrophenone
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.