missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoroindole, 97%

Manufacturer:  Alfa Aesar™ H56011MD

 View more versions of this product

Catalog No. AAH56011MD



Specifications

Specifications

4-Fluoroindole
30°C to 32°C
387-43-9
MFCD00055992
ZWKIJOPJWWZLDI-UHFFFAOYSA-N
4-fluoro-1H-indole
2774502
97%
90°C (0.4mmHg)
250mg
C8H6FN
1h-indole, 4-fluoro
C1=CC2=C(C=CN2)C(=C1)F
135.141
135.14

Chemical Identifiers

387-43-9
135.141
ZWKIJOPJWWZLDI-UHFFFAOYSA-N
2774502
C1=CC2=C(C=CN2)C(=C1)F
C8H6FN
MFCD00055992
1h-indole, 4-fluoro
4-fluoro-1H-indole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents