missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorocinnamic acid, 99%, ACROS Organics™

Manufacturer:  Acros Organics 207110050

 View more versions of this product

Catalog No. AC207110050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Fluorocinnamic acid
Glass bottle
207.0°C to 210.0°C
99%
98.5
99%
FC6H4CH=CHCO2H
09, II, 395
ISMMYAZSUSYVQG-ZZXKWVIFSA-N
(E)-3-(4-fluorophenyl)prop-2-enoic acid
1530234
Crystalline Solid
Authentic
White
5g
459-32-5
100.0
C9H7FO2
MFCD00004395
4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e
C1=CC(=CC=C1C=CC(=O)O)F
166.151
166.15
99%

Chemical Identifiers

459-32-5
166.151
ISMMYAZSUSYVQG-ZZXKWVIFSA-N
1530234
C1=CC(=CC=C1C=CC(=O)O)F
C9H7FO2
MFCD00004395
4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e
(E)-3-(4-fluorophenyl)prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 207-288-9

SDS Documents