missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorochalcone, 98%, ACROS Organics™

$62.24 - $62.24

Chemical Identifiers

CAS 1608-51-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C15H11FO
Formula Weight 226.25
MDL Number MFCD00443467
Synonym 1-Phenyl-3-(4-fluorophenyl)-2-propen-1-one
Chemical Name or Material 4-Fluorochalcone, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC358540050 SDS Thermo Fisher Scientific
AC358540050
5g Each for $62.24
Specifications Description & Specifications

Chemical Identifiers

1608-51-1
100.0
C15H11FO
MFCD00443467
4-Fluorochalcone, 98%
97.5
98%
226.25
1-Phenyl-3-(4-fluorophenyl)-2-propen-1-one
≥97.5%

Specifications

Yellow
5g
Authentic
100.0
98%
226.25
07, IV, 1659
4-Fluorochalcone, 98%
84° to 88°C
1608-51-1
97.5
Crystalline Powder
C15H11FO
MFCD00443467
1-Phenyl-3-(4-fluorophenyl)-2-propen-1-one
≥97.5%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.