Please login to your online account to display your discounted pricing

4-Fluorochalcone, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC358540050

 View more versions of this product

Catalog No. AC358540050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 226.25
Physical Form Crystalline Powder
Quantity 5g
Color Yellow
Melting Point 84° to 88°C
Beilstein 07, IV, 1659
CAS Max % 100.0
CAS 1608-51-1
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Assay Percent Range 98%
CAS Min % 97.5
Synonym 1-Phenyl-3-(4-fluorophenyl)-2-propen-1-one
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Fluorochalcone
MDL Number MFCD00443467
Name Note 98%
Molecular Formula C15H11FO

Specifications

Chemical Name or Material 4-Fluorochalcone
Name Note 98%
CAS 1608-51-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H11FO
Formula Weight 226.25
MDL Number MFCD00443467
Synonym 1-Phenyl-3-(4-fluorophenyl)-2-propen-1-one
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents