Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluorobutyrophenone, 97%

$142.24 - $521.34

Chemical Identifiers

Chemical Name or Material 4'-Fluorobutyrophenone
CAS 582-83-2
Molecular Formula C10H11FO
Formula Weight 166.2
MDL Number MFCD00017967
Synonym 1-(4-Fluorophenyl)-1-butanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2603403

  SDS
alfa aesar™
H2603403
1g Each for $142.24

AAH2603406

  SDS
alfa aesar™
H2603406
5g Each for $521.34
Description & Specifications

Specifications

Formula Weight 166.2
Chemical Name or Material 4'-Fluorobutyrophenone
Assay 97%
Molecular Formula C10H11FO
MDL Number MFCD00017967
CAS 582-83-2
Synonym 1-(4-Fluorophenyl)-1-butanone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 35° to 38°C
Quantity 1g

Applications
4′-fluorobutyrophenone derivatives is used in the preparation of its derivatives that includes haloperidol, haloanisone, triperidol, methylperidide, haloperidide and dipiperone.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.