Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluorobiphenyl-4-boronic acid, 98%

Chemical Identifiers

Chemical Name or Material 4'-Fluorobiphenyl-4-boronic acid
CAS 140369-67-1
Formula Weight 216.02
MDL Number MFCD08544387
Beilstein 8689987
Synonym 4-(4-Fluorophenyl)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6442103

  SDS
alfa aesar™
H6442103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6442106

  SDS
alfa aesar™
H6442106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64421MD

  SDS
alfa aesar™
H64421MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.02
Chemical Name or Material 4'-Fluorobiphenyl-4-boronic acid
Physical Form Solid
Assay 98%
MDL Number MFCD08544387
CAS 140369-67-1
Beilstein 8689987
Synonym 4-(4-Fluorophenyl)phenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C