Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzylurea, 98%

$70.95 - $210.88

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzylurea
CAS 76523-24-5
Molecular Formula C8H9FN2O
Formula Weight 168.17
MDL Number MFCD00085086
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1286503

 
alfa aesar™
A1286503
1g Each for $70.95

AAA1286506

 
alfa aesar™
A1286506
5g Each for $210.88
Description & Specifications

Specifications

Formula Weight 168.17
Chemical Name or Material 4-Fluorobenzylurea
Assay 98%
Molecular Formula C8H9FN2O
MDL Number MFCD00085086
CAS 76523-24-5
Melting Point 172° to 175°C
Quantity 1g