Please login to your online account to display your discounted pricing

4-Fluorobenzylamine, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  171950050

 View more versions of this product

Catalog No. AC171950050

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzylamine
Density 1.09g/mL
Color Clear colorless to light yellow
Quantity 5g
CAS 140-75-0
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Physical Form Liquid
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C7H8FN
Linear Formula FC6H4CH2NH2
Formula Weight 125.15

Specifications

Chemical Name or Material 4-Fluorobenzylamine
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C7H8FN
Linear Formula FC6H4CH2NH2
Formula Weight 125.15