Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Fluorobenzyl)triphenylphosphonium chloride, 98+%

$44.00 - $44.00

Chemical Identifiers

Chemical Name or Material (4-Fluorobenzyl)triphenylphosphonium chloride
CAS 3462-95-1
Molecular Formula C25H21ClFP
Formula Weight 406.87
MDL Number MFCD00031637
Beilstein 4075331
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1085206

  SDS
alfa aesar™
A1085206
5g Each for $44.00
Description & Specifications

Specifications

Formula Weight 406.87
Beilstein 4075331
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Chemical Name or Material (4-Fluorobenzyl)triphenylphosphonium chloride
CAS 3462-95-1
Molecular Formula C25H21ClFP
Assay ≥98%
Health Hazard 1 Warning
MDL Number MFCD00031637
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 310° to 312°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.