missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1221403

 View more versions of this product

Catalog No. AAL1221403



Specifications

Specifications

4-Fluorobenzyl isothiocyanate
124°C to 125°C (6mmHg)
2740-88-7
MFCD00041115
2085915
LPVNPJMEWWUFHD-UHFFFAOYSA-N
1-fluoro-4-(isothiocyanatomethyl)benzene
75963
97%
Irritating
1g
C8H6FNS
UN2922
4-fluorobenzyl isothiocyanate, 1-fluoro-4-isothiocyanatomethyl benzene, 4-fluorobenzylisothiocyanate, p-fluorobenzyl isothiocyanate, benzene, 1-fluoro-4-isothiocyanatomethyl, 4-fluorophenyl methanisothiocyanate, pubchem5031, acmc-20ami3, 4-fluorobenzyl-isothiocyanate, 4-isothiocyanatomethyl fluorobenzene
C1=CC(=CC=C1CN=C=S)F
167.201
167.2

Chemical Identifiers

2740-88-7
167.201
LPVNPJMEWWUFHD-UHFFFAOYSA-N
75963
C1=CC(=CC=C1CN=C=S)F
C8H6FNS
MFCD00041115
4-fluorobenzyl isothiocyanate, 1-fluoro-4-isothiocyanatomethyl benzene, 4-fluorobenzylisothiocyanate, p-fluorobenzyl isothiocyanate, benzene, 1-fluoro-4-isothiocyanatomethyl, 4-fluorophenyl methanisothiocyanate, pubchem5031, acmc-20ami3, 4-fluorobenzyl-isothiocyanate, 4-isothiocyanatomethyl fluorobenzene
1-fluoro-4-(isothiocyanatomethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents