Please login to your online account to display your discounted pricing

4-Fluorobenzyl isocyanate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC319300050

 View more versions of this product

Catalog No. AC319300050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 151.15
Quantity 5g
Color Colorless
Density 1.186g/mL
CAS Max % 100.0
Linear Formula FC6H4CHNCO
Molecular Formula C8H6FNO
Infrared Spectrum Authentic
Additional Information Readily forms solids from hydrolysis
Chemical Name or Material 4-Fluorobenzyl isocyanate
Assay Percent Range 98%
MDL Number MFCD00673062
CAS 132740-43-3
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.5040 to 1.5060 (20°C, 589nm)

Specifications

Chemical Name or Material 4-Fluorobenzyl isocyanate
CAS 132740-43-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H6FNO
Linear Formula FC6H4CHNCO
Formula Weight 151.15
MDL Number MFCD00673062
Accessories

GHS Signal Word: Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

SDS & Safety Documents