Learn More
4-Fluorobenzyl chloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 119431000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Fluorobenzyl chloride | |
352-11-4 | |
100.0 | |
181°C | |
Authentic | |
Glass bottle | |
1.5120 to 1.514 | |
MFCD00000913 | |
1.207 | |
IZXWCDITFDNEBY-UHFFFAOYSA-N | |
1-(chloromethyl)-4-fluorobenzene | |
9602 | |
98% |
98% | |
97.5 | |
1.2070g/mL | |
72°C | |
97.5% min. (GC) | |
C7H6ClF | |
FC6H4CH2Cl | |
100mL | |
4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene | |
C1=CC(=CC=C1CCl)F | |
144.58 | |
144.58 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 206-516-4
TSCA : TSCA
RUO â Research Use Only