missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzyl bromide, 97%

Manufacturer:  Alfa Aesar™ A1020830

 View more versions of this product

Catalog No. AAA1020830



Specifications

Specifications

4-Fluorobenzyl bromide
Pungent
250g
C7H6BrF
UN3265
4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
C1=CC(=CC=C1CBr)F
189.027
189.03
66°C (150°F)
84°C to 85°C (15mmHg)
459-46-1
MFCD00000359
636507
NVNPLEPBDPJYRZ-UHFFFAOYSA-N
1-(bromomethyl)-4-fluorobenzene
68021
97%

Chemical Identifiers

459-46-1
189.027
NVNPLEPBDPJYRZ-UHFFFAOYSA-N
68021
C1=CC(=CC=C1CBr)F
C7H6BrF
MFCD00000359
4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
1-(bromomethyl)-4-fluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents