missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluorobenzyl Alcohol 98.0+%, TCI America™
Supplier: TCI America F025125G
Specifications
4-Fluorobenzyl Alcohol | |
White-Yellow | |
C7H7FO | |
25 g | |
GEZMEIHVFSWOCA-UHFFFAOYSA-N | |
(4-fluorophenyl)methanol | |
68022 | |
≥98.0% (GC) |
459-56-3 | |
99°C | |
MFCD00004651 | |
4-fluorobenzyl alcohol, 4-fluorophenyl methanol, benzenemethanol, 4-fluoro, p-fluorobenzyl alcohol, 4-fluorobenzylalcohol, 4-fluorobenzylic alcohol, ccris 5115, 4-fluorobenzoic alcohol, 4-fluoro benzyl alcohol, 4-fluorophenyl methan-1-ol | |
C1=CC(=CC=C1CO)F | |
126.13 | |
126.13 | |
Crystalline Lumps |
Chemical Identifiers
459-56-3 | |
126.13 | |
GEZMEIHVFSWOCA-UHFFFAOYSA-N | |
68022 | |
C1=CC(=CC=C1CO)F |
C7H7FO | |
MFCD00004651 | |
4-fluorobenzyl alcohol, 4-fluorophenyl methanol, benzenemethanol, 4-fluoro, p-fluorobenzyl alcohol, 4-fluorobenzylalcohol, 4-fluorobenzylic alcohol, ccris 5115, 4-fluorobenzoic alcohol, 4-fluoro benzyl alcohol, 4-fluorophenyl methan-1-ol | |
(4-fluorophenyl)methanol |
Safety and Handling
TSCA : No