Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzyl alcohol, 97%

$46.80 - $227.81

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzyl alcohol
CAS 459-56-3
Molecular Formula C7H7FO
Formula Weight 126.13
MDL Number MFCD00004651
Beilstein 1446930
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1233606

 
alfa aesar™
A1233606
5g Each for $46.80

AAA1233614

 
alfa aesar™
A1233614
25g Each for $147.92

AAA1233618

 
alfa aesar™
A1233618
50g Each for $227.81
Description & Specifications

Specifications

Formula Weight 126.13
Chemical Name or Material 4-Fluorobenzyl alcohol
Assay 97%
Molecular Formula C7H7FO
MDL Number MFCD00004651
CAS 459-56-3
Beilstein 1446930
Health Hazard 1 Warning
Flash Point 90°C (194°F)
Odor Characteristic
Boiling Point 206°C
Melting Point 22° to 24°C
Quantity 5g

Applications
4-Fluorobenzyl alcohol is used in the preparation of many compounds including amides. It is also used as an pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acid anhydrides.

Safety and Handling

Warning