missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzyl alcohol, 97%

Manufacturer:  Alfa Aesar™ A1233614

 View more versions of this product

Catalog No. AAA1233614


 

Description

Description

Applications
4-Fluorobenzyl alcohol is used in the preparation of many compounds including amides. It is also used as an pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acid anhydrides.
Specifications

Specifications

4-Fluorobenzyl alcohol
Characteristic
22°C to 24°C
459-56-3
C7H7FO
1446930
C1=CC(=CC=C1CO)F
126.13
126.13
90°C (194°F)
206°C
25g
97%
MFCD00004651
GEZMEIHVFSWOCA-UHFFFAOYSA-N
(4-fluorophenyl)methanol
68022

Chemical Identifiers

459-56-3
126.13
GEZMEIHVFSWOCA-UHFFFAOYSA-N
C7H7FO
MFCD00004651
68022
Safety and Handling

Safety and Handling

Warning

SDS Documents