Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzoylacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzoylacetonitrile
CAS 4640-67-9
Formula Weight 163.15
MDL Number MFCD00662062
UN Number UN3439
Synonym 4-Fluorophenacyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6198806

  SDS
alfa aesar™
H6198806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6198803

  SDS
alfa aesar™
H6198803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 84° to 88°C
Formula Weight 163.15
MDL Number MFCD00662062
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 4-Fluorophenacyl cyanide
Color Off-white to pale yellow
Physical Form Solid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
UN Number UN3439
CAS 4640-67-9
Assay 97%
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Quantity 5g
Chemical Name or Material 4-Fluorobenzoylacetonitrile

Applications
Used as pharmaceutical intermediates.

Solubility
Soluble in dimethylformamide, dimethyl sulfoxide.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.