missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzoylacetonitrile, 97%

Manufacturer:  Alfa Aesar™ H6198803

 View more versions of this product

Catalog No. AAH6198803



Description

Description

Applications
Used as pharmaceutical intermediates.

Solubility
Soluble in dimethylformamide, dimethyl sulfoxide.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

4-Fluorobenzoylacetonitrile
84°C to 88°C
4640-67-9
MFCD00662062
3-4-fluoro-phenyl-3-oxo-propionitrile
C1=CC(=CC=C1C(=O)CC#N)F
163.151
163.15
97%
White to Yellow
1g
C9H6FNO
UN3439
LOJBBLDAJBJVBZ-UHFFFAOYSA-N
3-(4-fluorophenyl)-3-oxopropanenitrile
2783172
Solid

Chemical Identifiers

4640-67-9
163.151
LOJBBLDAJBJVBZ-UHFFFAOYSA-N
2783172
C1=CC(=CC=C1C(=O)CC#N)F
C9H6FNO
MFCD00662062
3-4-fluoro-phenyl-3-oxo-propionitrile
3-(4-fluorophenyl)-3-oxopropanenitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents