Learn More
4-Fluorobenzoyl chloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 152950250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Fluorobenzoyl chloride | |
403-43-0 | |
100.0 | |
82.0°C (20.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5299 to 1.5319 | |
MFCD00000684 | |
09, I, 137 | |
benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride | |
CZKLEJHVLCMVQR-UHFFFAOYSA-N | |
4-fluorobenzoyl chloride | |
67879 | |
98% |
98% | |
97.5 | |
1.3400g/mL | |
82°C | |
97.5% min. (GC) | |
C7H4ClFO | |
FC6H4COCl | |
25mL | |
1.34 | |
Solubility in water: decomposes | |
C1=CC(=CC=C1C(=O)Cl)F | |
158.56 | |
158.56 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 206-961-4
TSCA : TSCA
RUO â Research Use Only