Please login to your online account to display your discounted pricing

4-Fluorobenzotrifluoride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzotrifluoride
Name Note 97%
CAS 402-44-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4F4
Linear Formula FC6H4CF3
Formula Weight 164.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC187390050

  SDS
thermo fisher scientific
187390050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC187390250

  SDS
thermo fisher scientific
187390250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 164.1
Quantity 5g
Color Clear colorless
EINECS Number 206-944-1
TSCA TSCA
Density 1.2900g/mL
Beilstein 05, III, 680
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Fluorobenzotrifluoride
Name Note 97%
Assay Percent Range 97%
MDL Number MFCD00000399
CAS 402-44-8
Infrared Spectrum Authentic
Synonym α,α,α,4-Tetrafluorotoluene
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.4004 to 1.4024 (20°C, 589nm)
Molecular Formula C7H4F4
Linear Formula FC6H4CF3
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.