missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorobenzophenone 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 119381000

 View more versions of this product

Catalog No. AC119381000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Fluorobenzophenone
Authentic
159.0°C to 161.0°C (13.0mmHg)
43.0°C to 50.0°C
98%
97.5
98%
FC6H4COC6H5
07, I, 226
OGTSHGYHILFRHD-UHFFFAOYSA-N
(4-fluorophenyl)-phenylmethanone
67663
Crystalline Powder
>110°C
Glass bottle
Beige
100g
345-83-5
100.0
C13H9FO
MFCD00000352
4'-fluorobenzophenone, 4-fluoro-benzophenone, 4-fluorobenzophenone, 4-fluorophenyl phenyl ketone, 4-fluorophenyl phenyl methanone, 4-fluorophenyl-phenylmethanone, benzophenone, 4-fluoro, methanone, 4-fluorophenyl phenyl, p-fluorobenzophenone, pubchem3308
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)F
200.21
200.21
98%

Chemical Identifiers

345-83-5
200.21
OGTSHGYHILFRHD-UHFFFAOYSA-N
67663
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)F
C13H9FO
MFCD00000352
4'-fluorobenzophenone, 4-fluoro-benzophenone, 4-fluorobenzophenone, 4-fluorophenyl phenyl ketone, 4-fluorophenyl phenyl methanone, 4-fluorophenyl-phenylmethanone, benzophenone, 4-fluoro, methanone, 4-fluorophenyl phenyl, p-fluorobenzophenone, pubchem3308
(4-fluorophenyl)-phenylmethanone
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 206-463-7

SDS Documents