missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorobenzophenone 98%, ACROS Organics™

Chemical Identifiers

CAS 345-83-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C13H9FO
Linear Formula FC6H4COC6H5
Formula Weight 200.21
MDL Number MFCD00000352
Synonym (4-Fluorophenyl)(phenyl)methanone
Chemical Name or Material 4-Fluorobenzophenone, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC119381000 SDS Thermo Fisher Scientific
119381000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

345-83-5
100.0
C13H9FO
200.21
(4-Fluorophenyl)(phenyl)methanone
≥97.5%
97.5
98%
FC6H4COC6H5
MFCD00000352
4-Fluorobenzophenone, 98%

Specifications

Light beige
100g
Authentic
100.0
Crystalline Powder
C13H9FO
200.21
07, I, 226
4-Fluorobenzophenone, 98%
43° to 50°C
345-83-5
97.5
Glass bottle
98%
FC6H4COC6H5
MFCD00000352
(4-Fluorophenyl)(phenyl)methanone
≥97.5%