missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorobenzonitrile, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 160810250

 View more versions of this product

Catalog No. AC160810250



Specifications

Specifications

4-Fluorobenzonitrile
Authentic
188°C (750.0mmHg)
33°C to 36°C
99%
98.5
98.5% min. (GC)
FC6H4CN
09, I, 138
AEKVBBNGWBBYLL-UHFFFAOYSA-N
4-fluorobenzonitrile
14517
Crystalline Low Melting Solid
65°C
Glass bottle
White
25g
1194-02-1
100.0
C7H4FN
MFCD00001812
1-cyano-4-fluorobenzene
C1=CC(=CC=C1C#N)F
121.11
121.11
99%

Chemical Identifiers

1194-02-1
121.11
AEKVBBNGWBBYLL-UHFFFAOYSA-N
14517
C1=CC(=CC=C1C#N)F
C7H4FN
MFCD00001812
1-cyano-4-fluorobenzene
4-fluorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fre

GHS Signal Word: Warning

EINECSNumber : 214-784-9

RTECSNumber : DI4368500

TSCA : TSCA

SDS Documents