missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluorobenzoic hydrazide, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438100050
Specifications
4-Fluorobenzoic hydrazide | |
Glass bottle | |
161°C to 164°C | |
456-06-4 | |
100.0 | |
C7H7FN2O | |
UIVXXFYJRYVRKJ-UHFFFAOYSA-N | |
4-fluorobenzohydrazide | |
9972 | |
Crystalline Powder |
Authentic | |
Brown to Orange | |
5g | |
95.0 | |
95% min. (HPLC) | |
4-fluoro benzoyl hydrazine, 4-fluoro-benzoic acid hydrazide, 4-fluorobenzenecarbohydrazide, 4-fluorobenzhydrazide, 4-fluorobenzoic acid hydrazide, 4-fluorobenzoic hydrazide, benzoic acid, p-fluoro-, hydrazide, hydrazine, p-fluorobenzoyl, p-fluorobenzoic acid hydrazide, p-fluorobenzoylhydrazine | |
C1=CC(=CC=C1C(=O)NN)F | |
154.14 | |
154.14 | |
96% |
Chemical Identifiers
456-06-4 | |
154.14 | |
4-fluoro benzoyl hydrazine, 4-fluoro-benzoic acid hydrazide, 4-fluorobenzenecarbohydrazide, 4-fluorobenzhydrazide, 4-fluorobenzoic acid hydrazide, 4-fluorobenzoic hydrazide, benzoic acid, p-fluoro-, hydrazide, hydrazine, p-fluorobenzoyl, p-fluorobenzoic acid hydrazide, p-fluorobenzoylhydrazine | |
4-fluorobenzohydrazide |
C7H7FN2O | |
UIVXXFYJRYVRKJ-UHFFFAOYSA-N | |
9972 | |
C1=CC(=CC=C1C(=O)NN)F |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning