missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorobenzoic hydrazide, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438100010

 View more versions of this product

Catalog No. AC438100010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Fluorobenzoic hydrazide
Glass Bottle
161°C to 164°C
456-06-4
100.0
C7H7FN2O
UIVXXFYJRYVRKJ-UHFFFAOYSA-N
4-fluorobenzohydrazide
9972
Crystalline Powder
Authentic
Brown to Orange
1g
95.0
95% min. (HPLC)
4-fluoro benzoyl hydrazine, 4-fluoro-benzoic acid hydrazide, 4-fluorobenzenecarbohydrazide, 4-fluorobenzhydrazide, 4-fluorobenzoic acid hydrazide, 4-fluorobenzoic hydrazide, benzoic acid, p-fluoro-, hydrazide, hydrazine, p-fluorobenzoyl, p-fluorobenzoic acid hydrazide, p-fluorobenzoylhydrazine
C1=CC(=CC=C1C(=O)NN)F
154.14
154.14
96%

Chemical Identifiers

456-06-4
154.14
4-fluoro benzoyl hydrazine, 4-fluoro-benzoic acid hydrazide, 4-fluorobenzenecarbohydrazide, 4-fluorobenzhydrazide, 4-fluorobenzoic acid hydrazide, 4-fluorobenzoic hydrazide, benzoic acid, p-fluoro-, hydrazide, hydrazine, p-fluorobenzoyl, p-fluorobenzoic acid hydrazide, p-fluorobenzoylhydrazine
4-fluorobenzohydrazide
C7H7FN2O
UIVXXFYJRYVRKJ-UHFFFAOYSA-N
9972
C1=CC(=CC=C1C(=O)NN)F
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

GHS Signal Word: Warning

SDS Documents
Product Certifications