Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzoic acid, 98%

$39.03 - $432.45

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzoic acid
CAS 456-22-4
Molecular Formula C7H5FO2
Formula Weight 140.11
MDL Number MFCD00002530
Beilstein 1906922
Merck Index 14,4171
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1412914

 
alfa aesar™
A1412914
25g Each for $39.03

AAA1412922

 
alfa aesar™
A1412922
100g Each for $101.26

AAA1412936

 
alfa aesar™
A1412936
500g Each for $432.45
Description & Specifications

Specifications

Formula Weight 140.11
Assay 98%
CAS 456-22-4
Beilstein 1906922
Health Hazard 1 Warning
Chemical Name or Material 4-Fluorobenzoic acid
TSCA TSCA
Molecular Formula C7H5FO2
MDL Number MFCD00002530
Merck Index 14,4171
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 183° to 187°C
Quantity 25g

Applications
4-Fluorobenzoic acid is used as an intermediate for agrochemicals and pharmaceuticals.

Solubility
Soluble in alcohol, hot water, methanol, and ether. Very slightly soluble in cold water.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.