Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzhydrazide, 97%

$262.18 - $262.18

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzhydrazide
CAS 456-06-4
Molecular Formula C7H7FN2O
Formula Weight 154.14
MDL Number MFCD00060562
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5349614

  SDS
alfa aesar™
H5349614
25g Each for $262.18
Description & Specifications

Specifications

Formula Weight 154.14
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C7H7FN2O
CAS 456-06-4
Quantity 25g
MDL Number MFCD00060562
Health Hazard 1 Warning
Chemical Name or Material 4-Fluorobenzhydrazide
Melting Point 160° to 162°C

Applications
Used in the stnthesis of fluorinated poly (1, 3, 4-oxadiazole-ether-imide). N-(3,5-dinitrobenzoyl)-N′-(4-fluorobenzoyl)-hydrazine with 4-Fluorobenzhydrazide as solvent in NMP.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.