missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzhydrazide, 97%

Manufacturer:  Alfa Aesar™ H5349614

 View more versions of this product

Catalog No. AAH5349614



Description

Description

Applications
Used in the stnthesis of fluorinated poly (1, 3, 4-oxadiazole-ether-imide). N-(3,5-dinitrobenzoyl)-N′-(4-fluorobenzoyl)-hydrazine with 4-Fluorobenzhydrazide as solvent in NMP.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

4-Fluorobenzhydrazide
25g
C7H7FN2O
4-fluoro benzoyl hydrazine
C1=CC(=CC=C1C(=O)NN)F
154.144
154.14
160°C to 162°C
456-06-4
MFCD00060562
UIVXXFYJRYVRKJ-UHFFFAOYSA-N
4-fluorobenzohydrazide
9972
97%

Chemical Identifiers

456-06-4
154.144
UIVXXFYJRYVRKJ-UHFFFAOYSA-N
9972
C1=CC(=CC=C1C(=O)NN)F
C7H7FN2O
MFCD00060562
4-fluoro benzoyl hydrazine
4-fluorobenzohydrazide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents