Please login to your online account to display your discounted pricing

4-Fluorobenzenesulfonyl isocyanate, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzenesulfonyl isocyanate
Name Note 95%
CAS 3895-25-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7 H4 F N O3 S
Linear Formula FC6H4SO2NCO
Formula Weight 201.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC319285000

  SDS
thermo fisher scientific
319285000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Low Melting Solid
Formula Weight 201.09
Quantity 500mg
Color White
Melting Point 32° to 39°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 4-Fluorobenzenesulfonyl isocyanate
Name Note 95%
Assay Percent Range 95%
Molecular Formula C7 H4 F N O3 S
Linear Formula FC6H4SO2NCO
MDL Number MFCD00673073
CAS 3895-25-8
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.