missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorobenzenesulfonyl isocyanate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 319285000

 View more versions of this product

Catalog No. AC319285000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Fluorobenzenesulfonyl isocyanate
32° to 39°C
95%
Authentic
100.0
Crystalline Low Melting Solid
95%
FC6H4SO2NCO
MFCD00673073
White
500mg
3895-25-8
94.0
Glass bottle
≥94%
C7 H4 F N O3 S
201.09

Chemical Identifiers

3895-25-8
100.0
C7 H4 F N O3 S
201.09
4-Fluorobenzenesulfonyl isocyanate, 95%
94.0
95%
FC6H4SO2NCO
MFCD00673073
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning