missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorobenzenesulfonyl Chloride, TCI America™

Manufacturer:  TCI America F020225G

 View more versions of this product

Catalog No. F020225G



Specifications

Specifications

4-Fluorobenzenesulfonyl Chloride
White
25g
C6H4ClFO2S
3261
BFXHJFKKRGVUMU-UHFFFAOYSA-N
4-fluorobenzenesulfonyl chloride
9588
Crystal-Lump at 20°C
112°C
35°C
349-88-2
MFCD00007438
4-fluorobenzene-1-sulfonyl chloride, p-fluorobenzenesulfonyl chloride, 4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro, 4-fluorophenylsulfonyl chloride, 4-fluorobenzenesulfonic acid chloride, 4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, p-fluoro, 4-fluorobenzensulfonyl chloride, 4-fluoro-benzenesulfonyl chloride
C1=CC(=CC=C1F)S(=O)(=O)Cl
194.604
194.60
≥97.0% (GC,T)

Chemical Identifiers

349-88-2
194.604
BFXHJFKKRGVUMU-UHFFFAOYSA-N
9588
C1=CC(=CC=C1F)S(=O)(=O)Cl
C6H4ClFO2S
MFCD00007438
4-fluorobenzene-1-sulfonyl chloride, p-fluorobenzenesulfonyl chloride, 4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro, 4-fluorophenylsulfonyl chloride, 4-fluorobenzenesulfonic acid chloride, 4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, p-fluoro, 4-fluorobenzensulfonyl chloride, 4-fluoro-benzenesulfonyl chloride
4-fluorobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

TSCA : Yes

Documents