missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzenesulfonamide, 98+%

Manufacturer:  Alfa Aesar™ H5583403

 View more versions of this product

Catalog No. AAH5583403



Description

Description

Applications
4-Fluorobenzenesulfonamide, is used to produce bis-(4-fluoro-benzenesulfonyl)-amine with 4-fluoro-benzenesulfonyl chloride at temperature of 55 - 60. This reaction will need reagent OH-.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

4-Fluorobenzenesulfonamide
1g
C6H6FNO2S
UN2811
It is soluble in water.
C1=CC(=CC=C1F)S(=O)(=O)N
175.177
175.18
124°C to 127°C
402-46-0
MFCD00025384
4-flourobenzenesulfonamide
LFLSATHZMYYIAQ-UHFFFAOYSA-N
4-fluorobenzenesulfonamide
120231
≥98%

Chemical Identifiers

402-46-0
175.177
LFLSATHZMYYIAQ-UHFFFAOYSA-N
120231
C1=CC(=CC=C1F)S(=O)(=O)N
C6H6FNO2S
MFCD00025384
4-flourobenzenesulfonamide
4-fluorobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 206-946-2

RTECSNumber : DB2630000

TSCA : No

Recommended Storage : Ambient temperatures

Documents