missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluorobenzamidine hydrochloride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6157003
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-Fluorobenzamidine hydrochloride is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
4-Fluorobenzamidine hydrochloride | |
C7H8ClFN2 | |
1 g | |
JQDATBKJKUWNGA-UHFFFAOYSA-N | |
4-fluorobenzenecarboximidamide;hydrochloride | |
12456160 | |
95% |
456-14-4 | |
MFCD04114421 | |
4-fluorobenzamidine hydrochloride, 4-fluorobenzimidamide hydrochloride, 4-fluorobenzamidine hcl, 4-fluoro-benzamidine hydrochloride, 4-fluorobenzenecarboximidamide hydrochloride, 4-fluorobenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-fluoro-, monohydrochloride, 4-fluorobenzimidamidehydrochloride, 4-fluorobenzamidine hydrogen chloride, 4-fluoranylbenzenecarboximidamide hydrochloride | |
[H+].[Cl-].NC(=N)C1=CC=C(F)C=C1 | |
174.60 | |
174.6 | |
Solid |
Chemical Identifiers
456-14-4 | |
174.60 | |
JQDATBKJKUWNGA-UHFFFAOYSA-N | |
12456160 | |
[H+].[Cl-].NC(=N)C1=CC=C(F)C=C1 |
C7H8ClFN2 | |
MFCD04114421 | |
4-fluorobenzamidine hydrochloride, 4-fluorobenzimidamide hydrochloride, 4-fluorobenzamidine hcl, 4-fluoro-benzamidine hydrochloride, 4-fluorobenzenecarboximidamide hydrochloride, 4-fluorobenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-fluoro-, monohydrochloride, 4-fluorobenzimidamidehydrochloride, 4-fluorobenzamidine hydrogen chloride, 4-fluoranylbenzenecarboximidamide hydrochloride | |
4-fluorobenzenecarboximidamide;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures