missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzamidine hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H6157006

 View more versions of this product

Catalog No. AAH6157006



Description

Description

Applications
4-Fluorobenzamidine hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-Fluorobenzamidine hydrochloride
456-14-4
MFCD04114421
JQDATBKJKUWNGA-UHFFFAOYSA-N
4-fluorobenzenecarboximidamide;hydrochloride
12456160
Solid
5g
C7H8ClFN2
4-fluoranylbenzenecarboximidamide hydrochloride
C1=CC(=CC=C1C(=N)N)F.Cl
174.603
174.6
95%

Chemical Identifiers

456-14-4
174.603
JQDATBKJKUWNGA-UHFFFAOYSA-N
12456160
C1=CC(=CC=C1C(=N)N)F.Cl
C7H8ClFN2
MFCD04114421
4-fluoranylbenzenecarboximidamide hydrochloride
4-fluorobenzenecarboximidamide;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents