Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzamide, 98%

$30.45 - $244.09

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzamide
CAS 824-75-9
Molecular Formula C7H6FNO
Formula Weight 139.13
MDL Number MFCD00007992
Beilstein 1859744
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0683106

  SDS
alfa aesar™
L0683106
5g Each for $30.45

AAL0683114

  SDS
alfa aesar™
L0683114
25g Each for $83.18

AAL0683122

  SDS
alfa aesar™
L0683122
100g Each for $244.09
Description & Specifications

Specifications

Formula Weight 139.13
MDL Number MFCD00007992
Health Hazard 1 Warning
Chemical Name or Material 4-Fluorobenzamide
Molecular Formula C7H6FNO
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 154° to 157°C
Beilstein 1859744
RTECS Number CV4956666
Quantity 5g
CAS 824-75-9
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.