Learn More
4-Fluorobenzamide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0683114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Fluorobenzamide | |
154°C to 157°C | |
MFCD00007992 | |
1859744 | |
VNDHYTGVCGVETQ-UHFFFAOYSA-N | |
4-fluorobenzamide | |
71572 | |
98% |
824-75-9 | |
C7H6FNO | |
25 g | |
p-fluorobenzamide, benzamide, 4-fluoro, benzamide, p-fluoro, p-fluorobenzoic acid amide, 4-fluoro-benzamide, unii-yb79f3xb1g, yb79f3xb1g, 4fluorobenzamide, 4-fluorobenzoamide, pubchem3407 | |
C1=CC(=CC=C1C(=O)N)F | |
139.129 | |
139.13 |
Chemical Identifiers
824-75-9 | |
139.129 | |
VNDHYTGVCGVETQ-UHFFFAOYSA-N | |
71572 | |
C1=CC(=CC=C1C(=O)N)F |
C7H6FNO | |
MFCD00007992 | |
p-fluorobenzamide, benzamide, 4-fluoro, benzamide, p-fluoro, p-fluorobenzoic acid amide, 4-fluoro-benzamide, unii-yb79f3xb1g, yb79f3xb1g, 4fluorobenzamide, 4-fluorobenzoamide, pubchem3407 | |
4-fluorobenzamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 212-535-9
RTECSNumber : CV4956666
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only